Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATATGAATGCATAATTTTATAGA[C/T]GTAAGCATTGTATTTAATTATAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605545 | ||||||||||||||||||||||||||||||||
Literature Links: |
CD163 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CD163 - CD163 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004244.5 | Intron | NP_004235.4 | ||||
NM_203416.3 | Intron | NP_981961.2 | ||||
XM_005253528.4 | Intron | XP_005253585.1 | ||||
XM_005253529.4 | Intron | XP_005253586.1 | ||||
XM_017020231.1 | Intron | XP_016875720.1 |
Set Membership: |
HapMap |