Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTATTTGGGGCCTGTGGTTTAAC[C/T]TTTTGGATCACTCTGTGATATGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616009 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPS7A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COPS7A - COP9 signalosome subunit 7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164093.1 | Intron | NP_001157565.1 | ||||
NM_001164094.1 | Intron | NP_001157566.1 | ||||
NM_001164095.2 | Intron | NP_001157567.1 | ||||
NM_016319.3 | Intron | NP_057403.1 | ||||
XM_005253694.2 | Intron | XP_005253751.1 |