Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
54 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606095 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPIH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPIH - peptidylprolyl isomerase H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006347.3 | 923 | Intron | NP_006338.1 | |||
XM_005270362.1 | 923 | Silent Mutation | GCA,GCG | A,A 113 | XP_005270419.1 | |
XM_005270366.3 | 923 | Silent Mutation | GCA,GCG | A,A 82 | XP_005270423.1 | |
XM_011540508.1 | 923 | Intron | XP_011538810.1 | |||
XM_017000057.1 | 923 | Silent Mutation | GCA,GCG | A,A 113 | XP_016855546.1 | |
XM_017000058.1 | 923 | Silent Mutation | GCA,GCG | A,A 70 | XP_016855547.1 | |
XM_017000059.1 | 923 | Silent Mutation | GCA,GCG | A,A 70 | XP_016855548.1 |
Set Membership: |
HapMap |