Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCACTTCAGCCCTCTGAGCCTCT[C/T]TTCATGAGGGTAGAGGTCGGGCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 162340 MIM: 616144 | ||||||||||||||||||||
Literature Links: |
NMB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NMB - neuromedin B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021077.3 | 12 | UTR 5 | NP_066563.2 | |||
NM_205858.1 | 12 | UTR 5 | NP_995580.1 | |||
XM_017022239.1 | 12 | UTR 5 | XP_016877728.1 |
WDR73 - WD repeat domain 73 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |