Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 189964 MIM: 601558 | ||||||||||||||||||||||||||
Literature Links: |
GTF2E2 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
GTF2E2 - general transcription factor IIE subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002095.4 | 813 | Missense Mutation | AAA,GAA | K,E 234 | NP_002086.1 | |
XM_011544509.2 | 813 | Intron | XP_011542811.1 | |||
XM_011544510.1 | 813 | Intron | XP_011542812.1 | |||
XM_017013363.1 | 813 | Missense Mutation | AAA,GAA | K,E 234 | XP_016868852.1 | |
XM_017013364.1 | 813 | Missense Mutation | AAA,GAA | K,E 234 | XP_016868853.1 |
RBPMS - RNA binding protein with multiple splicing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |