Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACTTTTTTTCCCTGAATATTTT[G/T]TGGAGCCAGCTGCATTATAGGATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJA4 - DnaJ heat shock protein family (Hsp40) member A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130182.1 | Intron | NP_001123654.1 | ||||
NM_001130183.1 | Intron | NP_001123655.1 | ||||
NM_018602.3 | Intron | NP_061072.3 | ||||
XM_005254535.3 | Intron | XP_005254592.1 | ||||
XM_006720601.1 | Intron | XP_006720664.1 |
Set Membership: |
HapMap |