Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTTCCAGACCAGGAGGAAGTGC[C/G]TCTCTTCTTTCCGCTGCACCTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPL - MPL proto-oncogene, thrombopoietin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005373.2 | 350 | Missense Mutation | CCT,CGT | P,R 102 | NP_005364.1 | |
XM_017001320.1 | 350 | Missense Mutation | CCT,CGT | P,R 159 | XP_016856809.1 |