Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTTGCAGTAAGTGTGAGTCAGG[A/C]CCCCCTCCCAGGGCTGTGGCCCTCG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607139 MIM: 608460 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCA PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
FANCA - Fanconi anemia complementation group A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000135.2 | 5093 | UTR 3 | NP_000126.2 | |||
NM_001018112.1 | 5093 | Intron | NP_001018122.1 | |||
NM_001286167.1 | 5093 | UTR 3 | NP_001273096.1 | |||
XM_005256294.4 | 5093 | Intron | XP_005256351.1 | |||
XM_011522945.2 | 5093 | Intron | XP_011521247.1 | |||
XM_011522946.2 | 5093 | Intron | XP_011521248.1 | |||
XM_011522947.2 | 5093 | Intron | XP_011521249.1 | |||
XM_011522948.2 | 5093 | Intron | XP_011521250.1 | |||
XM_017023044.1 | 5093 | Intron | XP_016878533.1 | |||
XM_017023045.1 | 5093 | Intron | XP_016878534.1 | |||
XM_017023046.1 | 5093 | Intron | XP_016878535.1 |
ZNF276 - zinc finger protein 276 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113525.1 | 5093 | Intron | NP_001106997.1 | |||
NM_152287.3 | 5093 | Intron | NP_689500.2 | |||
XM_005256324.3 | 5093 | Intron | XP_005256381.1 | |||
XM_005256328.3 | 5093 | Intron | XP_005256385.1 | |||
XM_017023889.1 | 5093 | Intron | XP_016879378.1 | |||
XM_017023890.1 | 5093 | Intron | XP_016879379.1 |
Set Membership: |
HapMap |