Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 148065 MIM: 148030 MIM: 148020 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KRT13 - keratin 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRT15 - keratin 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002275.3 | 1505 | Missense Mutation | AAG,AGG | K,R 416 | NP_002266.2 | |
XM_011524784.2 | 1505 | Missense Mutation | AAG,AGG | K,R 423 | XP_011523086.1 | |
XM_017024614.1 | 1505 | Missense Mutation | AAG,AGG | K,R 423 | XP_016880103.1 |
KRT19 - keratin 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6510 - microRNA 6510 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |