Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCACGATGTCTTTCTCAGATGCA[T/C]CCTATTAGGGAAAAGAACACAAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138130 | ||||||||||||||||||||
Literature Links: |
GLUD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLUD1 - glutamate dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318900.1 | 2127 | Missense Mutation | NP_001305829.1 | |||
NM_001318901.1 | 2127 | Missense Mutation | NP_001305830.1 | |||
NM_001318902.1 | 2127 | Missense Mutation | NP_001305831.1 | |||
NM_001318904.1 | 2127 | Missense Mutation | NP_001305833.1 | |||
NM_001318905.1 | 2127 | Missense Mutation | NP_001305834.1 | |||
NM_001318906.1 | 2127 | Missense Mutation | NP_001305835.1 | |||
NM_005271.4 | 2127 | Missense Mutation | NP_005262.1 |