Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 306480 MIM: 300747 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4767 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR4767 - microRNA 4767 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PUDP - pseudouridine 5'-phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135565.1 | 118 | Intron | NP_001129037.1 | |||
NM_001178135.1 | 118 | Intron | NP_001171606.1 | |||
NM_001178136.1 | 118 | Intron | NP_001171607.1 | |||
NM_012080.4 | 118 | Intron | NP_036212.3 |
STS - steroid sulfatase (microsomal), isozyme S | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000351.5 | 118 | Intron | NP_000342.2 | |||
NM_001320750.1 | 118 | Intron | NP_001307679.1 | |||
NM_001320751.1 | 118 | UTR 5 | NP_001307680.1 | |||
NM_001320752.1 | 118 | UTR 5 | NP_001307681.1 | |||
NM_001320753.1 | 118 | Intron | NP_001307682.1 | |||
NM_001320754.1 | 118 | Intron | NP_001307683.1 |
Set Membership: |
HapMap |