Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607273 MIM: 614960 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLCN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FLCN - folliculin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144606.5 | 4076 | Intron | NP_653207.1 | |||
NM_144997.5 | 4076 | UTR 3 | NP_659434.2 | |||
XM_011523714.2 | 4076 | UTR 3 | XP_011522016.1 | |||
XM_011523716.1 | 4076 | UTR 3 | XP_011522018.1 | |||
XM_011523718.2 | 4076 | UTR 3 | XP_011522020.1 | |||
XM_011523719.2 | 4076 | Intron | XP_011522021.1 | |||
XM_011523721.2 | 4076 | UTR 3 | XP_011522023.1 | |||
XM_017024304.1 | 4076 | UTR 3 | XP_016879793.1 | |||
XM_017024305.1 | 4076 | UTR 3 | XP_016879794.1 | |||
XM_017024306.1 | 4076 | Intron | XP_016879795.1 | |||
XM_017024307.1 | 4076 | Intron | XP_016879796.1 | |||
XM_017024308.1 | 4076 | Intron | XP_016879797.1 | |||
XM_017024309.1 | 4076 | UTR 3 | XP_016879798.1 |
PLD6 - phospholipase D family member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |