Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605191 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTAF1 - B-TFIID TATA-box binding protein associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003972.2 | Intron | NP_003963.1 | ||||
XM_011540326.1 | Intron | XP_011538628.1 | ||||
XM_011540327.2 | Intron | XP_011538629.1 | ||||
XM_011540328.2 | Intron | XP_011538630.1 | ||||
XM_011540329.2 | Intron | XP_011538631.1 | ||||
XM_017016877.1 | Intron | XP_016872366.1 | ||||
XM_017016878.1 | Intron | XP_016872367.1 | ||||
XM_017016879.1 | Intron | XP_016872368.1 | ||||
XM_017016880.1 | Intron | XP_016872369.1 | ||||
XM_017016881.1 | Intron | XP_016872370.1 |