Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAAAGAACAGGTCCATAAGTAA[C/T]AGAAGGATGATGAAACAGCTCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614406 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130950 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100130950 - uncharacterized LOC100130950 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCIMP - SLP adaptor and CSK interacting membrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271842.1 | Intron | NP_001258771.1 | ||||
NM_001319190.1 | Intron | NP_001306119.1 | ||||
NM_207103.3 | Intron | NP_996986.1 | ||||
XM_017024615.1 | Intron | XP_016880104.1 |