Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613584 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH1L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALDH1L2 - aldehyde dehydrogenase 1 family member L2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034173.3 | 5866 | UTR 3 | NP_001029345.2 | |||
XM_011537986.2 | 5866 | UTR 3 | XP_011536288.1 | |||
XM_011537988.2 | 5866 | Intron | XP_011536290.1 | |||
XM_011537989.2 | 5866 | Intron | XP_011536291.1 | |||
XM_017018889.1 | 5866 | UTR 3 | XP_016874378.1 | |||
XM_017018890.1 | 5866 | Intron | XP_016874379.1 |
Set Membership: |
HapMap |