Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCGCTGCAGTCGCCCGCCTGGCC[G/T]GCCTTCCCGTTCCTCCGCCTCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191840 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf55 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf55 - chromosome 10 open reading frame 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001791.2 | 1006 | UTR 3 | NP_001001791.2 |
PLAU - plasminogen activator, urokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145031.2 | 1006 | Intron | NP_001138503.1 | |||
NM_001319191.1 | 1006 | Intron | NP_001306120.1 | |||
NM_002658.4 | 1006 | Intron | NP_002649.1 | |||
XM_011539866.2 | 1006 | Intron | XP_011538168.1 |