Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAAGGATGTTCAACAAAGTGTC[A/C]GGGGTCTCTTTGAGCCACTGCTTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604044 MIM: 616731 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929657 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101929657 - uncharacterized LOC101929657 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEK3 - NIMA related kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146099.1 | 1561 | Silent Mutation | NP_001139571.1 | |||
NM_002498.2 | 1561 | Silent Mutation | NP_002489.1 | |||
NM_152720.2 | 1561 | Silent Mutation | NP_689933.1 |
NEK5 - NIMA related kinase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |