Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCTTGCTTTCTTCATGCCCTCC[C/G]CACTGCTCCTGCCACCTTTAGATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606627 MIM: 603707 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAAM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAAM2 - dishevelled associated activator of morphogenesis 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201427.1 | 3801 | UTR 3 | NP_001188356.1 | |||
NM_015345.3 | 3801 | UTR 3 | NP_056160.2 | |||
XM_006715039.3 | 3801 | UTR 3 | XP_006715102.3 | |||
XM_006715040.3 | 3801 | UTR 3 | XP_006715103.3 | |||
XM_006715042.2 | 3801 | UTR 3 | XP_006715105.1 | |||
XM_006715043.2 | 3801 | UTR 3 | XP_006715106.1 | |||
XM_006715045.3 | 3801 | UTR 3 | XP_006715108.1 | |||
XM_006715046.3 | 3801 | UTR 3 | XP_006715109.1 | |||
XM_017010630.1 | 3801 | UTR 3 | XP_016866119.1 |
LOC100505635 - uncharacterized LOC100505635 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOCS1 - molybdenum cofactor synthesis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001075098.3 | 3801 | UTR 3 | NP_001068566.1 | |||
NM_005943.5 | 3801 | UTR 3 | NP_005934.2 | |||
XM_011514632.2 | 3801 | Intron | XP_011512934.1 | |||
XM_011514633.2 | 3801 | UTR 3 | XP_011512935.1 | |||
XM_011514634.2 | 3801 | Intron | XP_011512936.1 | |||
XM_017010888.1 | 3801 | UTR 3 | XP_016866377.1 |