Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611501 MIM: 603427 MIM: 603657 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMTA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAMTA1 - calmodulin binding transcription activator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PER3 - period circadian clock 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAMP3 - vesicle associated membrane protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004781.3 | Intron | NP_004772.1 | ||||
XM_017002841.1 | Intron | XP_016858330.1 |