Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603334 | ||||||||||||||||||||
Literature Links: |
DNAH3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAH3 - dynein axonemal heavy chain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017539.2 | Intron | NP_060009.1 | ||||
XM_006721062.1 | Intron | XP_006721125.1 | ||||
XM_011545882.2 | Intron | XP_011544184.1 | ||||
XM_011545883.1 | Intron | XP_011544185.1 | ||||
XM_011545885.2 | Intron | XP_011544187.1 | ||||
XM_011545886.2 | Intron | XP_011544188.1 | ||||
XM_011545888.2 | Intron | XP_011544190.1 | ||||
XM_011545889.2 | Intron | XP_011544191.1 | ||||
XM_017023426.1 | Intron | XP_016878915.1 | ||||
XM_017023427.1 | Intron | XP_016878916.1 | ||||
XM_017023428.1 | Intron | XP_016878917.1 | ||||
XM_017023429.1 | Intron | XP_016878918.1 | ||||
XM_017023430.1 | Intron | XP_016878919.1 | ||||
XM_017023431.1 | Intron | XP_016878920.1 | ||||
XM_017023432.1 | Intron | XP_016878921.1 |