Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600183 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf105 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C17orf105 - chromosome 17 open reading frame 105 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136483.1 | Intron | NP_001129955.1 | ||||
XM_011524633.1 | Intron | XP_011522935.1 | ||||
XM_011524634.2 | Intron | XP_011522936.1 | ||||
XM_011524635.1 | Intron | XP_011522937.1 | ||||
XM_011524636.1 | Intron | XP_011522938.1 |
DUSP3 - dual specificity phosphatase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |