Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP42 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
USP42 - ubiquitin specific peptidase 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032172.2 | 218 | Silent Mutation | ACA,ACG | T,T 51 | NP_115548.1 | |
XM_005249883.4 | 218 | Silent Mutation | ACA,ACG | T,T 51 | XP_005249940.1 | |
XM_006715790.1 | 218 | Silent Mutation | ACA,ACG | T,T 51 | XP_006715853.1 | |
XM_006715791.2 | 218 | Silent Mutation | ACA,ACG | T,T 51 | XP_006715854.1 | |
XM_011515573.1 | 218 | Silent Mutation | ACA,ACG | T,T 51 | XP_011513875.1 | |
XM_011515574.1 | 218 | UTR 5 | XP_011513876.1 | |||
XM_011515577.1 | 218 | Intron | XP_011513879.1 | |||
XM_011515578.1 | 218 | UTR 5 | XP_011513880.1 |
Set Membership: |
HapMap |