Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600019 MIM: 615047 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYB561 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CYB561 - cytochrome b561 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017916.1 | 939 | UTR 3 | NP_001017916.1 | |||
NM_001017917.1 | 939 | UTR 3 | NP_001017917.1 | |||
NM_001915.3 | 939 | UTR 3 | NP_001906.3 | |||
XM_005257091.1 | 939 | UTR 3 | XP_005257148.1 | |||
XM_017024260.1 | 939 | UTR 3 | XP_016879749.1 | |||
XM_017024261.1 | 939 | UTR 3 | XP_016879750.1 | |||
XM_017024262.1 | 939 | UTR 3 | XP_016879751.1 |
TANC2 - tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |