Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGTTCATGGACTTGAGAACCCC[A/G]GAGTGGGAACAGCAGGTGCGGATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610334 MIM: 610385 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375566 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105375566 - uncharacterized LOC105375566 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM176A - transmembrane protein 176A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM176B - transmembrane protein 176B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101311.1 | 981 | UTR 3 | NP_001094781.1 | |||
NM_001101312.1 | 981 | UTR 3 | NP_001094782.1 | |||
NM_001101314.1 | 981 | UTR 3 | NP_001094784.1 | |||
NM_014020.3 | 981 | UTR 3 | NP_054739.3 | |||
XM_006715933.3 | 981 | UTR 3 | XP_006715996.2 |