Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602843 MIM: 610642 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGDIB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGDIB - Rho GDP dissociation inhibitor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001175.6 | Intron | NP_001166.3 | ||||
NM_001321420.1 | Intron | NP_001308349.1 | ||||
NM_001321421.1 | Intron | NP_001308350.1 | ||||
NM_001321422.1 | Intron | NP_001308351.1 | ||||
NM_001321423.1 | Intron | NP_001308352.1 |
ERP27 - endoplasmic reticulum protein 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |