Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601717 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PCP2 - Purkinje cell protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STXBP2 - syntaxin binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127396.2 | 535 | Silent Mutation | CGC,CGT | R,R 162 | NP_001120868.1 | |
NM_001272034.1 | 535 | Silent Mutation | CGC,CGT | R,R 176 | NP_001258963.1 | |
NM_006949.3 | 535 | Silent Mutation | CGC,CGT | R,R 165 | NP_008880.2 | |
XM_011528210.1 | 535 | Silent Mutation | CGC,CGT | R,R 165 | XP_011526512.1 | |
XM_011528212.2 | 535 | Silent Mutation | CGC,CGT | R,R 165 | XP_011526514.1 |
Set Membership: |
HapMap |