Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCGTTTCCTCAGTGTAGTTTTA[A/T]GGCCGTTATTACACTTTGTACTTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300644 MIM: 300610 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMCX4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARMCX4 - armadillo repeat containing, X-linked 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GLA - galactosidase alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HNRNPH2 - heterogeneous nuclear ribonucleoprotein H2 (H') | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032393.2 | Intron | NP_001027565.1 | ||||
NM_019597.4 | Intron | NP_062543.1 |
RPL36A-HNRNPH2 - RPL36A-HNRNPH2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199973.1 | Intron | NP_001186902.1 | ||||
NM_001199974.1 | Intron | NP_001186903.1 |
Set Membership: |
HapMap |