Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300107 MIM: 300346 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BRS3 - bombesin receptor subtype 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HTATSF1 - HIV-1 Tat specific factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163280.1 | Intron | NP_001156752.1 | ||||
NM_014500.4 | Intron | NP_055315.2 | ||||
XM_005262404.3 | Intron | XP_005262461.1 |
Set Membership: |
HapMap |