Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300283 MIM: 300005 MIM: 300929 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRAK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IRAK1 - interleukin 1 receptor associated kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MECP2 - methyl-CpG binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110792.1 | 4231 | Intron | NP_001104262.1 | |||
NM_001316337.1 | 4231 | Intron | NP_001303266.1 | |||
NM_004992.3 | 4231 | UTR 3 | NP_004983.1 | |||
XM_005274682.4 | 4231 | UTR 3 | XP_005274739.1 | |||
XM_006724819.3 | 4231 | UTR 3 | XP_006724882.1 | |||
XM_011531166.2 | 4231 | UTR 3 | XP_011529468.1 | |||
XM_017029533.1 | 4231 | UTR 3 | XP_016885022.1 |
MIR718 - microRNA 718 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |