Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606985 MIM: 607108 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ELP4 - elongator acetyltransferase complex subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAX6 - paired box 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000280.4 | 4294 | UTR 3 | NP_000271.1 | |||
NM_001127612.1 | 4294 | UTR 3 | NP_001121084.1 | |||
NM_001258462.1 | 4294 | UTR 3 | NP_001245391.1 | |||
NM_001258463.1 | 4294 | UTR 3 | NP_001245392.1 | |||
NM_001258464.1 | 4294 | UTR 3 | NP_001245393.1 | |||
NM_001258465.1 | 4294 | UTR 3 | NP_001245394.1 | |||
NM_001310158.1 | 4294 | UTR 3 | NP_001297087.1 | |||
NM_001310159.1 | 4294 | Intron | NP_001297088.1 | |||
NM_001310160.1 | 4294 | UTR 3 | NP_001297089.1 | |||
NM_001310161.1 | 4294 | UTR 3 | NP_001297090.1 | |||
NM_001604.5 | 4294 | UTR 3 | NP_001595.2 |