Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610177 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AEN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AEN - apoptosis enhancing nuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022767.3 | 253 | Intron | NP_073604.3 | |||
XM_005254966.1 | 253 | Intron | XP_005255023.1 | |||
XM_005254967.1 | 253 | Intron | XP_005255024.1 | |||
XM_006720645.3 | 253 | Intron | XP_006720708.2 | |||
XM_011521905.2 | 253 | Intron | XP_011520207.1 | |||
XM_017022489.1 | 253 | UTR 5 | XP_016877978.1 | |||
XM_017022490.1 | 253 | Intron | XP_016877979.1 |
LINC01586 - long intergenic non-protein coding RNA 1586 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1179 - microRNA 1179 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3529 - microRNA 3529 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR7-2 - microRNA 7-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |