Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616637 | ||||||||||||||||||||
Literature Links: |
TBC1D16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBC1D16 - TBC1 domain family member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271844.1 | 6244 | UTR 3 | NP_001258773.1 | |||
NM_001271845.1 | 6244 | UTR 3 | NP_001258774.1 | |||
NM_001271846.1 | 6244 | Intron | NP_001258775.1 | |||
NM_019020.3 | 6244 | UTR 3 | NP_061893.2 | |||
XM_005257049.2 | 6244 | UTR 3 | XP_005257106.1 | |||
XM_005257050.3 | 6244 | UTR 3 | XP_005257107.1 | |||
XM_005257052.3 | 6244 | Intron | XP_005257109.1 | |||
XM_006721694.3 | 6244 | UTR 3 | XP_006721757.1 | |||
XM_006721695.3 | 6244 | Intron | XP_006721758.1 | |||
XM_011524322.2 | 6244 | UTR 3 | XP_011522624.1 | |||
XM_011524323.2 | 6244 | UTR 3 | XP_011522625.2 | |||
XM_011524324.2 | 6244 | UTR 3 | XP_011522626.1 | |||
XM_017024185.1 | 6244 | Intron | XP_016879674.1 | |||
XM_017024186.1 | 6244 | UTR 3 | XP_016879675.1 | |||
XM_017024187.1 | 6244 | UTR 3 | XP_016879676.1 | |||
XM_017024188.1 | 6244 | UTR 3 | XP_016879677.1 |