Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606086 MIM: 606134 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF5B - eukaryotic translation initiation factor 5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REV1 - REV1, DNA directed polymerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037872.2 | Intron | NP_001032961.1 | ||||
NM_001321454.1 | Intron | NP_001308383.1 | ||||
NM_001321455.1 | Intron | NP_001308384.1 | ||||
NM_001321458.1 | Intron | NP_001308387.1 | ||||
NM_001321459.1 | Intron | NP_001308388.1 | ||||
NM_001321460.1 | Intron | NP_001308389.1 | ||||
NM_016316.3 | Intron | NP_057400.1 | ||||
XM_011511341.2 | Intron | XP_011509643.1 | ||||
XM_017004309.1 | Intron | XP_016859798.1 | ||||
XM_017004310.1 | Intron | XP_016859799.1 | ||||
XM_017004311.1 | Intron | XP_016859800.1 | ||||
XM_017004312.1 | Intron | XP_016859801.1 | ||||
XM_017004313.1 | Intron | XP_016859802.1 | ||||
XM_017004314.1 | Intron | XP_016859803.1 | ||||
XM_017004315.1 | Intron | XP_016859804.1 |