Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173490 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDGFRA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PDGFRA - platelet derived growth factor receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006206.4 | Intron | NP_006197.1 | ||||
XM_005265743.1 | Intron | XP_005265800.1 | ||||
XM_006714039.3 | Intron | XP_006714102.1 | ||||
XM_006714041.3 | Intron | XP_006714104.1 | ||||
XM_011534385.2 | Intron | XP_011532687.1 | ||||
XM_017008280.1 | Intron | XP_016863769.1 | ||||
XM_017008281.1 | Intron | XP_016863770.1 | ||||
XM_017008282.1 | Intron | XP_016863771.1 |
Set Membership: |
HapMap |