Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
87 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603886 MIM: 606494 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARTN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARTN - artemin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136215.1 | 838 | Missense Mutation | CAG,CGG | Q,R 19 | NP_001129687.1 | |
NM_057090.2 | 838 | Missense Mutation | CAG,CGG | Q,R 19 | NP_476431.2 | |
NM_057091.2 | 838 | Missense Mutation | CAG,CGG | Q,R 19 | NP_476432.2 |
ST3GAL3 - ST3 beta-galactoside alpha-2,3-sialyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |