Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160777 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO5A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MYO5A - myosin VA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000259.3 | 9874 | UTR 3 | NP_000250.3 | |||
NM_001142495.1 | 9874 | UTR 3 | NP_001135967.1 | |||
XM_005254397.3 | 9874 | Intron | XP_005254454.1 | |||
XM_011521606.2 | 9874 | UTR 3 | XP_011519908.2 | |||
XM_011521607.2 | 9874 | Intron | XP_011519909.2 | |||
XM_011521608.2 | 9874 | Intron | XP_011519910.2 | |||
XM_011521609.2 | 9874 | Intron | XP_011519911.2 | |||
XM_011521610.2 | 9874 | Intron | XP_011519912.2 | |||
XM_011521611.2 | 9874 | Intron | XP_011519913.2 | |||
XM_011521612.2 | 9874 | Intron | XP_011519914.2 | |||
XM_017022227.1 | 9874 | Intron | XP_016877716.1 |
Set Membership: |
HapMap |