Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGAATGTTCACAGCAAGATCA[C/T]CCTTAGTGGGTAAGCGGGGGCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL25 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLHL25 - kelch like family member 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022480.3 | Intron | NP_071925.2 |
LOC101929679 - uncharacterized LOC101929679 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1276 - microRNA 1276 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |