Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610858 MIM: 605399 | ||||||||||||||||||||
Literature Links: |
C14orf166 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C14orf166 - chromosome 14 open reading frame 166 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016039.2 | 4622 | Intron | NP_057123.1 |
NID2 - nidogen 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007361.3 | 4622 | UTR 3 | NP_031387.3 | |||
XM_005267405.4 | 4622 | UTR 3 | XP_005267462.1 | |||
XM_005267406.4 | 4622 | UTR 3 | XP_005267463.1 | |||
XM_005267407.4 | 4622 | UTR 3 | XP_005267464.1 |