Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600322 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNAP25 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SNAP25 - synaptosome associated protein 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322902.1 | Intron | NP_001309831.1 | ||||
NM_001322903.1 | Intron | NP_001309832.1 | ||||
NM_001322904.1 | Intron | NP_001309833.1 | ||||
NM_001322905.1 | Intron | NP_001309834.1 | ||||
NM_001322906.1 | Intron | NP_001309835.1 | ||||
NM_001322907.1 | Intron | NP_001309836.1 | ||||
NM_001322908.1 | Intron | NP_001309837.1 | ||||
NM_001322909.1 | Intron | NP_001309838.1 | ||||
NM_001322910.1 | Intron | NP_001309839.1 | ||||
NM_003081.4 | Intron | NP_003072.2 | ||||
NM_130811.3 | Intron | NP_570824.1 | ||||
XM_005260808.4 | Intron | XP_005260865.1 | ||||
XM_017028021.1 | Intron | XP_016883510.1 | ||||
XM_017028022.1 | Intron | XP_016883511.1 | ||||
XM_017028023.1 | Intron | XP_016883512.1 |
Set Membership: |
HapMap Validated |