Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613209 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM181 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM181 - transmembrane protein 181 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020823.1 | Intron | NP_065874.1 | ||||
XM_005267074.3 | Intron | XP_005267131.1 | ||||
XM_011535999.1 | Intron | XP_011534301.1 | ||||
XM_011536000.1 | Intron | XP_011534302.1 | ||||
XM_011536001.2 | Intron | XP_011534303.1 | ||||
XM_011536002.2 | Intron | XP_011534304.1 | ||||
XM_011536003.2 | Intron | XP_011534305.1 |
Set Membership: |
HapMap |