Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125950 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf76 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C2orf76 - chromosome 2 open reading frame 76 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017927.3 | 201 | Intron | NP_001017927.2 | |||
NM_001322329.1 | 201 | Intron | NP_001309258.1 | |||
NM_001322330.1 | 201 | Intron | NP_001309259.1 | |||
NM_001322331.1 | 201 | Intron | NP_001309260.1 | |||
NM_001322332.1 | 201 | Intron | NP_001309261.1 | |||
XM_017003354.1 | 201 | Intron | XP_016858843.1 |
DBI - diazepam binding inhibitor, acyl-CoA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079862.2 | 201 | Intron | NP_001073331.1 | |||
NM_001079863.1 | 201 | Intron | NP_001073332.1 | |||
NM_001178017.1 | 201 | Silent Mutation | CGA,CGG | R,R 39 | NP_001171488.1 | |
NM_001178041.2 | 201 | Silent Mutation | CGA,CGG | R,R 20 | NP_001171512.1 | |
NM_001178042.2 | 201 | Intron | NP_001171513.1 | |||
NM_001178043.2 | 201 | Intron | NP_001171514.1 | |||
NM_001282633.1 | 201 | Intron | NP_001269562.1 | |||
NM_001282634.1 | 201 | Intron | NP_001269563.1 | |||
NM_001282635.1 | 201 | Intron | NP_001269564.1 | |||
NM_001282636.1 | 201 | Intron | NP_001269565.1 | |||
NM_020548.7 | 201 | Intron | NP_065438.1 |
Set Membership: |
HapMap |