Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300889 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3H12B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZC3H12B - zinc finger CCCH-type containing 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010888.3 | Intron | NP_001010888.3 | ||||
XM_011530939.2 | Intron | XP_011529241.1 | ||||
XM_017029478.1 | Intron | XP_016884967.1 | ||||
XM_017029479.1 | Intron | XP_016884968.1 | ||||
XM_017029480.1 | Intron | XP_016884969.1 | ||||
XM_017029481.1 | Intron | XP_016884970.1 | ||||
XM_017029482.1 | Intron | XP_016884971.1 | ||||
XM_017029483.1 | Intron | XP_016884972.1 | ||||
XM_017029484.1 | Intron | XP_016884973.1 |
Set Membership: |
HapMap |