Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602623 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARMC12 - armadillo repeat containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286574.1 | Intron | NP_001273503.1 | ||||
NM_001286576.1 | Intron | NP_001273505.1 | ||||
NM_145028.4 | Intron | NP_659465.2 | ||||
XM_011514381.2 | Intron | XP_011512683.1 | ||||
XM_011514382.2 | Intron | XP_011512684.1 | ||||
XM_017010435.1 | Intron | XP_016865924.1 |
FKBP5 - FK506 binding protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC285847 - uncharacterized LOC285847 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |