Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610124 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C3orf22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C3orf22 - chromosome 3 open reading frame 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152533.2 | Intron | NP_689746.1 | ||||
XM_011512455.2 | Intron | XP_011510757.1 | ||||
XM_011512457.2 | Intron | XP_011510759.1 | ||||
XM_017005751.1 | Intron | XP_016861240.1 | ||||
XM_017005752.1 | Intron | XP_016861241.1 | ||||
XM_017005753.1 | Intron | XP_016861242.1 |
CHST13 - carbohydrate sulfotransferase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152889.2 | Intron | NP_690849.1 |