Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604554 | ||||||||||||||||||||
Literature Links: |
HSF2BP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSF2BP - heat shock transcription factor 2 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007031.1 | Intron | NP_008962.1 | ||||
XM_011529443.2 | Intron | XP_011527745.2 | ||||
XM_011529445.2 | Intron | XP_011527747.2 | ||||
XM_011529446.2 | Intron | XP_011527748.1 | ||||
XM_011529447.2 | Intron | XP_011527749.1 | ||||
XM_017028267.1 | Intron | XP_016883756.1 | ||||
XM_017028268.1 | Intron | XP_016883757.1 | ||||
XM_017028269.1 | Intron | XP_016883758.1 | ||||
XM_017028270.1 | Intron | XP_016883759.1 | ||||
XM_017028271.1 | Intron | XP_016883760.1 | ||||
XM_017028272.1 | Intron | XP_016883761.1 |