Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147440 | ||||||||||||||||||||
Literature Links: |
IGF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGF1 - insulin like growth factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000618.4 | 3651 | UTR 3 | NP_000609.1 | |||
NM_001111283.2 | 3651 | UTR 3 | NP_001104753.1 | |||
NM_001111284.1 | 3651 | UTR 3 | NP_001104754.1 | |||
XM_017019259.1 | 3651 | Intron | XP_016874748.1 | |||
XM_017019260.1 | 3651 | Intron | XP_016874749.1 | |||
XM_017019261.1 | 3651 | Intron | XP_016874750.1 | |||
XM_017019262.1 | 3651 | UTR 3 | XP_016874751.1 | |||
XM_017019263.1 | 3651 | UTR 3 | XP_016874752.1 |