Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182144 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHMT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SHMT1 - serine hydroxymethyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281786.1 | Intron | NP_001268715.1 | ||||
NM_004169.4 | Intron | NP_004160.3 | ||||
NM_148918.2 | Intron | NP_683718.1 | ||||
XM_005256767.3 | Intron | XP_005256824.1 | ||||
XM_011523992.2 | Intron | XP_011522294.1 | ||||
XM_017024957.1 | Intron | XP_016880446.1 | ||||
XM_017024958.1 | Intron | XP_016880447.1 | ||||
XM_017024959.1 | Intron | XP_016880448.1 | ||||
XM_017024960.1 | Intron | XP_016880449.1 |
SMCR8 - Smith-Magenis syndrome chromosome region, candidate 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |