Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186760 | ||||||||||||||||||||||||||||||||
Literature Links: |
CD28 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CD28 - CD28 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243077.1 | Intron | NP_001230006.1 | ||||
NM_001243078.1 | Intron | NP_001230007.1 | ||||
NM_006139.3 | Intron | NP_006130.1 | ||||
XM_011512194.2 | Intron | XP_011510496.1 | ||||
XM_011512195.2 | Intron | XP_011510497.1 | ||||
XM_011512197.2 | Intron | XP_011510499.1 |
Set Membership: |
HapMap |