Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607680 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RCHY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RCHY1 - ring finger and CHY zinc finger domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009922.2 | 1685 | UTR 3 | NP_001009922.1 | |||
NM_001278536.1 | 1685 | UTR 3 | NP_001265465.1 | |||
NM_001278537.1 | 1685 | UTR 3 | NP_001265466.1 | |||
NM_001278538.1 | 1685 | UTR 3 | NP_001265467.1 | |||
NM_001278539.1 | 1685 | UTR 3 | NP_001265468.1 | |||
NM_015436.3 | 1685 | UTR 3 | NP_056251.2 | |||
XM_011531838.2 | 1685 | UTR 3 | XP_011530140.1 | |||
XM_011531839.2 | 1685 | UTR 3 | XP_011530141.1 |
Set Membership: |
HapMap |