Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604735 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
USP16 - ubiquitin specific peptidase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001992.1 | Intron | NP_001001992.1 | ||||
NM_001032410.1 | Intron | NP_001027582.1 | ||||
NM_006447.2 | Intron | NP_006438.1 | ||||
XM_017028257.1 | Intron | XP_016883746.1 | ||||
XM_017028258.1 | Intron | XP_016883747.1 | ||||
XM_017028259.1 | Intron | XP_016883748.1 | ||||
XM_017028260.1 | Intron | XP_016883749.1 | ||||
XM_017028261.1 | Intron | XP_016883750.1 |
Set Membership: |
HapMap |